Ac-Tyr-Val-Ala-Asp-Aldehyde (Pseudo Acid)
Supplier: BACHEM AMERICAS INC
4018830.0005 4018830.0025
H-8410.0005BAEA 535.98 CAD
H-8410.0005BA H-8410.0025BA
Ac-Tyr-Val-Ala-Asp-Aldehyde (Pseudo Acid)
Proteins and Peptides
See also the Asp derivatives N-1425 and N-1430, and Z-YVAD-FMK (N-1595).
Ac-YVAD-CHO is a potent, specific, reversible inhibitor of caspase-1 (ICE). It displayed a Ki of 0.2 ± 0.1 nM for recombinant human caspase-1.