omega-Conotoxin MVIIA
Supplier: BACHEM AMERICAS INC
4025279.0500 4025279.1000
H-8210.0500BAEA 1161.77 CAD
H-8210.0500BA H-8210.1000BA
omega-Conotoxin MVIIA
Proteins and Peptides
ω-Conotoxin MVIIA, originally isolated from the venom of the fish-hunting cone snail Conus magus, is a blocker of voltage-sensitive Ca²⁺ channels in neurons. The peptide has been used to identify different Ca²⁺ channel subtypes in amphibian brain.