Human Amylin Trifluoroacetate

Supplier: BACHEM AMERICAS INC

4030200.0500 4030200.1000
H-7905.0500BAEA 774.5 CAD
H-7905.0500BA H-7905.1000BA
Human Amylin Trifluoroacetate
Proteins and Peptides

The amyloidogenic peptide hormone amylin 1-37 (or Islet Amyloid Polypeptide, IAPP), KCNTATCATQRLANFLVHSSNNFGAILSSTNVGSNTY amide, has been isolated from the amyloid-rich pancreases of diabetic patients. IAPP forms fibrillar peptide deposits in the pancreatic islets of Langerhans, which may be related to death of the insulin-producing islet β-cells in type 2 diabetes mellitus.

Order Now

SPECIFICATIONS

Protein/peptide name Amylin Trifluoroacetate
Environmentally Preferable
Protein synonyms Islet Amyloid Polypeptide|IAPP|Amlintide
Species Human
Source
CAS No. 122384-88-7
MDL No. MFCD00081345
Sequence H-Lys-Cys-Asn-Thr-Ala-Thr-Cys-Ala-Thr-Gln-Arg-Leu-Ala-Asn-Phe-Leu-Val-His-Ser-Ser-Asn-Asn-Phe-Gly-Ala-Ile-Leu-Ser-Ser-Thr-Asn-Val-Gly-Ser-Asn-Thr-Tyr-NH₂
Molecular weight 3903.33
Molecular formula C₁₆₅H₂₆₁N₅₁O₅₅S₂
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR