H-Ser-Asp-Gly-Arg-Gly-OH
Supplier: BACHEM AMERICAS INC
4015321.0005 4015321.0025
H-7740.0005BAEA 743.63 CAD
H-7740.0005BA H-7740.0025BA
H-Ser-Asp-Gly-Arg-Gly-OH
Proteins and Peptides
SDGRG represents a scrambled RGD peptide, the inverted sequence of H-1345. Used as control peptide for RGDS and GRGDS.