Salmon (Des-Gly¹⁰,D-Ala⁶,Pro-NHEt⁹)-LHRH
Supplier: BACHEM AMERICAS INC
4013835.0001 4013835.0005
H-7525.0001BAEA 645.43 CAD
H-7525.0001BA H-7525.0005BA
Salmon (Des-Gly¹⁰,D-Ala⁶,Pro-NHEt⁹)-LHRH
Proteins and Peptides
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
Contrary to mammals and other classes of species, the sequences of GnRH I-III found in fish show a high diversity. H-4258, Lamprey GnRH III (peforelin) has found use in veterinary medicine and the sGnRH analog H-9205 in aquaculture.