Human Recombinant EGF
Supplier: BACHEM AMERICAS INC
4030572.0100 4030572.0500
H-7490.0100BAEA 284.04 CAD
H-7490.0100BA H-7490.0500BA
Human Recombinant EGF
Proteins and Peptides
approx. ED₅₀ ≤ 0.1 ng/mL Human EGF (urogastrone, nepidermin) is a polypeptide hormone which was first isolated from human urine. It is a potent mitogen for a variety of cultured cells, e.g. fibroblasts. In vivo, urogastrone may have a role in the maintenance of gastrointestinal homeostasis. Furthermore, EGF is used in wound healing.