Hirudin Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4014556.0001 4014556.0005
H-7420.0001BAEA 325.67 CAD
H-7420.0001BA H-7420.0005BA
Hirudin Trifluoroacetate
Proteins and Peptides
These peptides correspond to the C-terminus of the 65-residue polypeptide hirudin, a potent thrombin inhibitor found in the saliva of the leech Hirudo medicinalis. Treatment of hirudin with carboxypeptidase Y has shown that the last 5 amino acids are essential for its inhibitory activity.