H-Gly-Arg-Gly-Asp-Ser-Pro-Cys-OH Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4030599.0005
4030599.0025
H-7245.0005BAEA
583.42
CAD
H-7245.0005BA
H-7245.0025BA
H-Gly-Arg-Gly-Asp-Ser-Pro-Cys-OH Trifluoroacetate
Proteins and Peptides
The integrin-binding heptapeptideGRGDSPC can be easily linked to carriers or surfaces.