Cyclo(-Arg-Ala-Asp-D-Phe-Cys) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4070810.0001 4070810.0005
H-7232.0001BAEA 206.6 CAD
H-7232.0001BA H-7232.0005BA
Cyclo(-Arg-Ala-Asp-D-Phe-Cys) Trifluoroacetate
Proteins and Peptides
c(RADfC), control peptide for the αvβ3 integrin binding cyclic RGD peptide c(RGDfC) (H-7226).