Thymosin alpha₁ Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4031308.0001 4031308.0005
H-6945.0001BAEA 645.43 CAD
H-6945.0001BA H-6945.0005BA
Thymosin alpha₁ Trifluoroacetate
Proteins and Peptides
Thymosin α₁, originally isolated from calf thymus, acts as an immunomodulator. Additionally, the peptide showed antiviral, angiogenic, and wound-healing activities. Thymosin α₁ was shown to activate tumor-associated macrophages.