Bovine;Guinea pig;Human;Mouse;Rat GLP-1 (7-36) amide Trifluoroacetate
Supplier: BACHEM AMERICAS INC
For the long-acting GLP-1 analog liraglutide see H-6724.
GLP-1 (7-36) amide is secreted from the lower small intestine and shows a strong insulinotropic effect. GLP-1 (7-36) amide is considered as the most important incretin hormone. Its action is mediated by receptors expressed by the endocrine pancreatic B-cells. Considerable interest has focused on the development of this peptide as a therapeutic strategy for non-insulin-dependent (type 2 ) diabetes mellitus and associated neuropathy.