Maxadilan Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4067317.0500 4067317.1000
H-6734.0500BAEA 2565.81 CAD
H-6734.0500BA H-6734.1000BA
Maxadilan Trifluoroacetate
Proteins and Peptides
Maxadilan, isolated from the salivary gland of the sand fly Lutzomyia longipalpis, is a potent vasodilator. The peptide specifically and potently activates the mammalian PAC1 receptor, one of the three receptors for PACAP.