Human;Mouse;Rat Adropin (34-76) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4064212.0500 4064212.1000
H-6686.0500BAEA 3180.16 CAD
H-6686.0500BA H-6686.1000BA
Human;Mouse;Rat Adropin (34-76) Trifluoroacetate
Proteins and Peptides
Adropin is a secreted factor involved in energy homeostasis and lipid metabolism. It is encoded by the energy homeostasis associated gene (Enho) and is expressed in liver and brain. In diet-induced obesity (DIO) mice Adropin (34-76) attenuated hepatosteatosis and insulin reistance independently of adiposity or food intake. Additionally, adropin could be a regulator of endothelial function.