Enfuvirtide Acetate
Supplier: BACHEM AMERICAS INC
4062216.0001 4062216.0005
H-6684.0001BAEA 380.91 CAD
H-6684.0001BA H-6684.0005BA
Enfuvirtide Acetate
Proteins and Peptides
Antimicrobial peptides are produced by plants and most organisms throughout the animal kingdom including humans. Antimicrobial peptides protect against a broad range of infectious agents, as bacteria, fungi, and viruses. The amphibian skin is an especially rich source of antimicrobial peptides. See also the product families: Hepcidins LL-37 and Fragments Tuftsin and Analogs (subfamily).
Enfuvirtide is a 36 amino acid peptide corresponding to a region of gp41, the transmembrane subunit of HIV-1 envelope protein. It belongs to the therapeutic class of fusion inhibitors and acts by binding to gp41 and impeding the conformational changes in gp41 necessary for fusion of the virus with the cell. Offered under Bolar Exemption: All APIs that are sold for development of drug products still patent protected are offered under Bolar Exemption only. The following disclaimer applies: These products are offered and sold in small quantities only and solely for uses reasonably related to privileged trials and studies for obtaining marketing authorization required by law (Bolar Exemption). Bachem cannot be made liable for any infringement of intellectual property rights. It is the sole and only responsibility of the purchaser or user of these products to comply with the relevant national rules and regulations.