Eptifibatide Acetate
Supplier: BACHEM AMERICAS INC
4063191.0001 4063191.0005 4063191.0025
H-6654.0001BAEA 180.91 CAD
H-6654.0001BA H-6654.0005BA H-6654.0025BA
Eptifibatide Acetate
Proteins and Peptides
Eptifibatide, a RGD mimetic, is a glycoprotein (GP) IIb/IIIa receptor antagonist. Clinically, the peptide is used as an antiplatelet drug in combination with heparin and aspirin in the management of unstable angina and in patients undergoing coronary angioplasty and stenting procedures. CAS Number (eptifibatide acetate): 148031-34-9. Offered under Bolar Exemption: All APIs that are sold for development of drug products still patent protected are offered under Bolar Exemption only. The following disclaimer applies: These products are offered and sold in small quantities only and solely for uses reasonably related to privileged trials and studies for obtaining marketing authorization required by law (Bolar Exemption). Bachem cannot be made liable for any infringement of intellectual property rights. It is the sole and only responsibility of the purchaser or user of these products to comply with the relevant national rules and regulations.