(Des-Gly¹⁰,D-Trp³,D-Leu⁶,Pro-NHEt⁹)-LHRH Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4058815.0001 4058815.0005
H-6636.0001BAEA 570.65 CAD
H-6636.0001BA H-6636.0005BA
(Des-Gly¹⁰,D-Trp³,D-Leu⁶,Pro-NHEt⁹)-LHRH Trifluoroacetate
Proteins and Peptides
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
Impurity E of the Leuprorelin Ph. Eur. monograph.