Vapreotide Acetate
Supplier: BACHEM AMERICAS INC
4062833.0005 4062833.0025
H-6634.0005BAEA 180.91 CAD
H-6634.0005BA H-6634.0025BA
Vapreotide Acetate
Proteins and Peptides
Vapreotide is a potent octapeptide analog of somatostatin with selectivity for the somatostatin receptor subtypes SSTR-2 and SSTR-5. It is under investigation in the management of various disorders, including esophageal variceal bleeding, gastrointestinal and pancreatic fistulas, acromegaly, carcinoid tumors, and for the prevention of postoperative complications following pancreatic surgery.