Rat Preptin Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4050843.0500 4050843.1000
H-6572.0500BAEA 1032.67 CAD
H-6572.0500BA H-6572.1000BA
Rat Preptin Trifluoroacetate
Proteins and Peptides
Preptin is a 34-amino acid peptide hormone, which is co-secreted with insulin from the pancreatic β-cells in response to glucose stimulation. It is osteogenic in vitro and in vivo and may act in concert with the other β-cell hormones insulin and amylin to stimulate bone formation in hyperinsulinemic states such as obesity.