Mouse CRAMP Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4056438.0001 4056438.0005
H-6526.0001BAEA 1007.49 CAD
H-6526.0001BA H-6526.0005BA
Mouse CRAMP Trifluoroacetate
Proteins and Peptides
Antimicrobial peptides are produced by plants and most organisms throughout the animal kingdom including humans. Antimicrobial peptides protect against a broad range of infectious agents, as bacteria, fungi, and viruses. The amphibian skin is an especially rich source of antimicrobial peptides. See also the product families: Hepcidins LL-37 and Fragments Tuftsin and Analogs (subfamily).
CRAMP is a cathelin-related antimicrobial peptide expressed in the embryonic and adult mouse. Functional studies showed CRAMP to be a potent antibiotic against Gram-negative bacteria by inhibiting growth of a variety of bacterial strains and by permeabilizing the inner membrane of E.coli directly.