Compstatin Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4045832.0500 4045832.1000
H-6516.0500BAEA 529.57 CAD
H-6516.0500BA H-6516.1000BA
Compstatin Trifluoroacetate
Proteins and Peptides
Compstatin, ICVVQDWGHHRCT, is a 13-residue cyclic peptide that inhibits complement activation by binding to complement component C3.