Human Kisspeptin-54 Trifluoroacetate

Supplier: BACHEM AMERICAS INC

4054937.0500 4054937.1000
H-6514.0500BAEA 1549.03 CAD
H-6514.0500BA H-6514.1000BA
Human Kisspeptin-54 Trifluoroacetate
Proteins and Peptides

Kisspeptin-54, an endogenous ligand of the G protein-coupled receptor GPR54, is a key regulator of the hypothalamo-pituitary-gonadal (HPG) axis. It stimulates LH and FSH secretion. Kisspeptin-54 may represent a versatile tool for the manipulation of the HPG axis.

Order Now

SPECIFICATIONS

Protein/peptide name Kisspeptin-54 Trifluoroacetate
Environmentally Preferable
Protein synonyms Metastin|KiSS-1 (68-121)|Malignant Melanoma Metastasis-Suppressor KiSS-1 (68-121)
Species Human
Source
CAS No. 374683-24-6
Sequence H-Gly-Thr-Ser-Leu-Ser-Pro-Pro-Pro-Glu-Ser-Ser-Gly-Ser-Arg-Gln-Gln-Pro-Gly-Leu-Ser-Ala-Pro-His-Ser-Arg-Gln-Ile-Pro-Ala-Pro-Gln-Gly-Ala-Val-Leu-Val-Gln-Arg-Glu-Lys-Asp-Leu-Pro-Asn-Tyr-Asn-Trp-Asn-Ser-Phe-Gly-Leu-Arg-Phe-NH₂
Molecular weight 5857.51
Molecular formula C₂₅₈H₄₀₁N₇₉O₇₈
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR