beta-Amyloid (1-42)
Supplier: BACHEM AMERICAS INC
4045866.0500 4045866.1000
H-6466.0500BAEA 1445.62 CAD
H-6466.0500BA H-6466.1000BA
beta-Amyloid (1-42)
Proteins and Peptides
Compared to the inner salt H-1368, the HCl salt of Aβ42 aggregates more readily at pH 7.4.