Ac-Gly-Asp-Tyr-Ser-His-Cys-Ser-Pro-Leu-Arg-Tyr-Tyr-Pro-Trp-Trp-Lys-Cys-Thr-Tyr-Pro-Asp-Pro-Glu-Gly-Gly-Gly-NH₂ Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4030130.0500 4030130.1000
H-6442.0500BAEA 1032.67 CAD
H-6442.0500BA H-6442.1000BA
Ac-Gly-Asp-Tyr-Ser-His-Cys-Ser-Pro-Leu-Arg-Tyr-Tyr-Pro-Trp-Trp-Lys-Cys-Thr-Tyr-Pro-Asp-Pro-Glu-Gly-Gly-Gly-NH₂ Trifluoroacetate
Proteins and Peptides
See also E-2410, Suc-Pro-OH, the product familiy 'Antihypertensive Peptides' and the subfamily 'Bradykinin-Potentiating Peptides (BPP)'.
DX600, Ac-GDYSHCSPLRYYPWWKCTYPDPEGGG-amide is an ACE-2-specific peptide inhibitor with a Ki of 2.8 nM.