beta-Amyloid (1-46)
Supplier: BACHEM AMERICAS INC
4050409.0500 4050409.1000
H-6406.0500BAEA 8853.23 CAD
H-6406.0500BA H-6406.1000BA
beta-Amyloid (1-46)
Proteins and Peptides
Precursor of the secreted amyloid β-protein (1-40) and (1-42). The identification of amyloid-β-protein (1-46) led to the identification of a zeta-cleavage site between the known γ- and ε-cleavage sites within the transmembrane domain of amyloid-β precursor protein (APP).