(Des-Gly¹⁰,Leu⁶,Pro-NHEt⁹)-LHRH Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4033802.0001 4033802.0005
H-6402.0001BAEA 1176.33 CAD
H-6402.0001BA H-6402.0005BA
(Des-Gly¹⁰,Leu⁶,Pro-NHEt⁹)-LHRH Trifluoroacetate
Proteins and Peptides
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
Impurity C of the Leuprorelin Ph. Eur. monograph.