(D-Ser(tBu)⁶,Azagly¹⁰)-LHRH Acetate
Supplier: BACHEM AMERICAS INC
4038252.0005 4038252.0025
H-6395.0005BAEA 258.14 CAD
H-6395.0005BA H-6395.0025BA
(D-Ser(tBu)⁶,Azagly¹⁰)-LHRH Acetate
Proteins and Peptides
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
Goserelin acetate, a synthetic peptide analog of LHRH, is used in the treatment of prostate cancer, advanced breast cancer and in the management of endometriosis. CAS Number (Goserelin net): 65807-02-5.