Bovine;Human;Mouse;Rat (Ala¹³)-Apelin-13 Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4050032.0001 4050032.0005
H-6308.0001BAEA 309.81 CAD
H-6308.0001BA H-6308.0005BA
Bovine;Human;Mouse;Rat (Ala¹³)-Apelin-13 Trifluoroacetate
Proteins and Peptides
Specific anatagonist of Apelin-13 (H-4566). Systemic administration of (Ala¹³)-Apelin-13 (30 µg/kg) abolished the blood pressure-lowering effects of H-4566 (15 µg/kg) in hypertensive rats.