Human Acetyl-Heme-Binding Protein 1 (1-21) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4050026.0001
4050026.0005
H-6294.0001BAEA
903.62
CAD
H-6294.0001BA
H-6294.0005BA
Human Acetyl-Heme-Binding Protein 1 (1-21) Trifluoroacetate
Proteins and Peptides
F2L is an acetylated peptide corresponding to an N-terminal fragment of the human heme-binding protein. Ac-MLGMIKNSLFGSVETWPWQVL acts specifically through the FPR-like receptor (FPRL) 2 promoting calcium mobilization and chemotaxis in dendritic cells and monocytes. In neutrophils, F2L inhibited FPR and FPRL1-mediated signaling.