HIV-1 tat Protein (47-57) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4048761.0005 4048761.0025
H-6254.0005BAEA 693.21 CAD
H-6254.0005BA H-6254.0025BA
HIV-1 tat Protein (47-57) Trifluoroacetate
Proteins and Peptides
YGRKKRRQRRR, a cell penetrating peptide derived from the HIV-1 tat protein transduction domain (PTD), has been shown to promote the entry of nucleic acids into several cell types.
Sequence: H-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-OH
Salt: Trifluoroacetate