For-Met-Ala-Ser-OH
Supplier: BACHEM AMERICAS INC
4013146.0050 4013146.0250
H-6210.0050BAEA 258.14 CAD
H-6210.0050BA H-6210.0250BA
For-Met-Ala-Ser-OH
Proteins and Peptides
fMAS was used as substrate in a continuous assay for peptide deformylase (PDF).