(D-Trp⁶)-LHRH
Supplier: BACHEM AMERICAS INC
4039648.0005 4039648.0025
H-6150.0005BAEA 774.5 CAD
H-6150.0005BA H-6150.0025BA
(D-Trp⁶)-LHRH
Proteins and Peptides
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
A long-acting GnRH agonist. Pamoate is the bivalent anion of pamoic acid (embonic acid, 4,4·-methylene-bis-(3-hydroxy-2-naphthoic acid)).