Human;Mouse;Rat Oxyntomodulin Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4044527.0500 4044527.1000
H-6058.0500BAEA 1831.39 CAD
H-6058.0500BA H-6058.1000BA
Human;Mouse;Rat Oxyntomodulin Trifluoroacetate
Proteins and Peptides
For the long-acting GLP-1 analog liraglutide see H-6724.
Oxyntomodulin potently inhibits gastric acid secretion and pancreatic enzyme secretion when infused iv. Moreover, it was shown that intracerebroventricularly and into the hypothalamic paraventricular nucleus injected oxyntomodulin inhibits food intake in fasted and nonfasted animals potently and in a sustained manner.