Mouse PAR-4 (1-6) amide Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4035649.0005 4035649.0025
H-6054.0005BAEA 238.17 CAD
H-6054.0005BA H-6054.0025BA
Mouse PAR-4 (1-6) amide Trifluoroacetate
Proteins and Peptides
PAR peptides (TRAP peptides and thrombin receptor-like peptides) activate the proteinase-activated receptors PAR-1 to PAR-4.
Protease-activated receptor-4 activating peptide derived from murine PAR-4. GYPGKF-amide was able to cause rat platelet aggregation with an EC₅₀ value of 40 µM. Its effect on leucocyte rolling and adherence points at a role of PAR-4 in mediating proinflammatory processes.