H-Gly-Arg-Ala-Asp-Ser-Pro-OH Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4043208.0005 4043208.0025
H-6048.0005BAEA 413.58 CAD
H-6048.0005BA H-6048.0025BA
H-Gly-Arg-Ala-Asp-Ser-Pro-OH Trifluoroacetate
Proteins and Peptides
GRADSP is used as negative control peptide for GRGDSP (H-7630).