Mouse (Ala¹)-PAR-4 (1-6) amide Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4035529.0005
4035529.0025
H-6046.0005BAEA
849.55
CAD
H-6046.0005BA
H-6046.0025BA
Mouse (Ala¹)-PAR-4 (1-6) amide Trifluoroacetate
Proteins and Peptides
This peptide based on the proteolytically-revealed tethered ligand sequence of the murine proteinase-activated receptor-4 (PAR-4) was shown to cause platelet aggregation with an EC50 value of about 15 µM.
Sequence: H-Ala-Tyr-Pro-Gly-Lys-Phe-NH2
Salt: Trifluoroacetate