Dog;Mouse;Pig;Rat Peptide YY (3-36) Trifluoroacetate

Supplier: BACHEM AMERICAS INC

4044052.0500 4044052.1000
H-6042.0500BAEA 516.34 CAD
H-6042.0500BA H-6042.1000BA
Dog;Mouse;Pig;Rat Peptide YY (3-36) Trifluoroacetate
Proteins and Peptides

Peptide YY (3-36) and peptide YY are both synthesized by the gastrointestinal tract and released into the circulation after a meal. Peptide YY (3-36) is a Y₂ receptor subtype agonist, whereas peptide YY is non-selective for Y₁ and Y₂ receptor subtypes. It has been suggested that Y₁ and Y₂ receptor subtype binding affinities depend on their secondary and tertiary solution state structures.

Order Now

SPECIFICATIONS

Protein/peptide name Peptide YY (3-36) Trifluoroacetate
Environmentally Preferable
Protein synonyms PYY (3-36)
Species Pig,Dog,Rat,Mouse
Source
CAS No. 126339-09-1
Sequence H-Ala-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Ser-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Gln-Arg-Tyr-NH₂
Molecular weight 3980.41
Molecular formula C₁₇₆H₂₇₂N₅₂O₅₄
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR