Human Orphan GPCR SP9155 Agonist P518 Trifluoroacetate, Bachem
Supplier: BACHEM AMERICAS INC
4043081.0500 4043081.1000
H-5984.0500BAEA 981.04 CAD
H-5984.0500BA H-5984.1000BA
Human Orphan GPCR SP9155 Agonist P518 Trifluoroacetate, Bachem
Proteins and Peptides
P518 (human) is an RF-amide peptide ligand for SP9155, an orphan G-protein-coupled receptor with high homology to orexin, neuropeptide FF and CCK receptors. It has been shown to potently activate SP9155 with an EC₅₀ of 7 nM.