Human Myelin Oligodendrocyte Glycoprotein (35-55) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4042037.0500
4042037.1000
H-5912.0500BAEA
464.74
CAD
H-5912.0500BA
H-5912.1000BA
Human Myelin Oligodendrocyte Glycoprotein (35-55) Trifluoroacetate
Proteins and Peptides
In contrast to the encephalitogenic rat homolog, myelin oligodendrocyte glycoprotein (35-55) (human) does not appear to be a major T-cell epitope in multiple sclerosis propably due to its lower amount of helical structure formed when dissolved in micellar sodium dodecyl sulfate, a membrane mimicking solvent.