Human Urocortin II Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4040984.0500 4040984.1000
H-5852.0500BAEA 774.5 CAD
H-5852.0500BA H-5852.1000BA
Human Urocortin II Trifluoroacetate
Proteins and Peptides
Ucn II (human), a new member of the CRF family of neuropeptides, binds selectively to the type 2 CRF receptor (CRF-R2), with no appreciable activity on CRF-R1.