Aprotinin Acetate

Supplier: BACHEM AMERICAS INC

4035911.0005 4035911.0025
H-5834.0005BAEA 264.44 CAD
H-5834.0005BA H-5834.0025BA
Aprotinin Acetate
Proteins and Peptides

Aprotinin (BPTI), a widely used serine protease inhibitor. Bachem's aprotinin is a synthetic product and is not derived from bovine origin.


  • Disulfide bonds between Cys5 and Cys55/Cys14 and Cys38/Cys30 and Cys51

Order Now

SPECIFICATIONS

Protein/peptide name Aprotinin Acetate
Environmentally Preferable
Protein synonyms Pancreatic Trypsin Inhibitor|BPI|Basic Protease Inhibitor|BPTI
Species
Source
CAS No. 9087-70-1
Sequence H-Arg-Pro-Asp-Phe-Cys-Leu-Glu-Pro-Pro-Tyr-Thr-Gly-Pro-Cys-Lys-Ala-Arg-Ile-Ile-Arg-Tyr-Phe-Tyr-Asn-Ala-Lys-Ala-Gly-Leu-Cys-Gln-Thr-Phe-Val-Tyr-Gly-Gly-Cys-Arg-Ala-Lys-Arg-Asn-Asn-Phe-Lys-Ser-Ala-Glu-Asp-Cys-Met-Arg-Thr-Cys-Gly-Gly-Ala-OH
Molecular weight 6511.53
Molecular formula C₂₈₄H₄₃₂N₈₄O₇₉S₇
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR