Bovine;Human;Mouse;Rat Apelin-12
Supplier: BACHEM AMERICAS INC
4038636.0001 4038636.0005
H-5806.0001BAEA 219.6 CAD
H-5806.0001BA H-5806.0005BA
Bovine;Human;Mouse;Rat Apelin-12
Proteins and Peptides
Apelin-12, RPRLSHKGPMPF, is an endogenous ligand of the human orphan receptor APJ and has been shown to reduce arterial blood pressure without producing a change in the heart rate in anesthetized rats. Its vasoactive effect might be exerted by activation of endothelial nitric oxide synthase.