Human;Mouse;Bovine Thymopentin Acetate
Supplier: BACHEM AMERICAS INC
4002968.0025 4002968.0100
H-5805.0025BAEA 180.83 CAD
H-5805.0025BA H-5805.0100BA
Human;Mouse;Bovine Thymopentin Acetate
Proteins and Peptides
See also the product families 'Adjuvant Peptides' and 'Thymosins and Fragments'.
Thymopentin (TP5), an active fragment of thymopoietin (TP), reduces endocrine and behavioral responses to experimental stress, possibly by lowering plasma TP (pTP) levels. The immunomodulatory peptide suppresses proliferation and induces differentiation in HL-60 cells.