Human GLP-2 (1-33) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4039611.0500 4039611.1000
H-5662.0500BAEA 529.57 CAD
H-5662.0500BA H-5662.1000BA
Human GLP-2 (1-33) Trifluoroacetate
Proteins and Peptides
For the long-acting GLP-1 analog liraglutide see H-6724.
Like GLP-1, GLP-2 is secreted from enteroendocrine cells in a nutrient-dependent manner in both rodents and humans. Currently GLP-2 is used as a potential therapeutic agent for human subjects with a broad variety of intestinal diseases characterized by intestinal damage and insufficiency.