beta-Amyloid (1-42),Biotin

Supplier: BACHEM AMERICAS INC

4038795.0500 4038795.1000
H-5642.0500BAEA 1288.55 CAD
H-5642.0500BA H-5642.1000BA
beta-Amyloid (1-42),Biotin
Proteins and Peptides

Biotinyl-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA.
Sequence: Biotinyl-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH
Salt: Ammonium

Order Now

SPECIFICATIONS

Protein/peptide name beta-Amyloid (1-42)
Environmentally Preferable
Species
Source
Conjugation Biotin
Sequence Biotinyl-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA.
Molecular weight 4740.40
Molecular formula C₂₁₃H₃₂₅N₅₇O₆₂S₂
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR