Human Urocortin III Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4039202.0500 4039202.1000
H-5634.0500BAEA 1059.11 CAD
H-5634.0500BA H-5634.1000BA
Human Urocortin III Trifluoroacetate
Proteins and Peptides
Urocortin III (human) is a highly selective ligand for the CRF-II receptor. The amino acid sequence of UCN III corresponds to amino acids 3-40 of stresscopin (human) (H-5546).