beta-Amyloid (1-40)
Supplier: BACHEM AMERICAS INC
4038267.0500
4038267.1000
H-5568.0500BAEA
757.18
CAD
H-5568.0500BA
H-5568.1000BA
beta-Amyloid (1-40)
Proteins and Peptides
Compared to the TFA salt, the HCl salt of amyloid β-protein (1-40) easily forms β-structures in PBS within a few hours at 25°C. β-structure formation is essential for amyloid peptide toxicity.
Sequence: H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH
Salt: Hydrochloride