Human Stresscopin
Supplier: BACHEM AMERICAS INC
4037535.0500 4037535.1000
H-5546.0500BAEA 1588.74 CAD
H-5546.0500BA H-5546.1000BA
Human Stresscopin
Proteins and Peptides
Stresscopin is a specific ligand for the corticotropin-releasing hormone receptor type 2 (CRHR2). Transcripts are expressed in brain and most tissues analyzed. Intraperitoneal injections of stresscopin suppresses heat-induced edema formation in anesthetized rats. It also decreases food intake and exhibits an inhibitory effect on gastric emptying activity. This CRHR2 agonist might represent an endogenous ligand for maintaining homeostasis after stress.