Pig Neuromedin U-25 Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4031100.0500 4031100.1000
H-5510.0500BAEA 516.34 CAD
H-5510.0500BA H-5510.1000BA
Pig Neuromedin U-25 Trifluoroacetate
Proteins and Peptides
Neuromedin U-8 and neuromedin U-25 from porcine spinal cord are potent stimulators of rat uterus smooth muscle. Both peptides have also been reported to influence blood pressure in rats and dogs