OM99-2 Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4034503.0500
4034503.1000
H-5108.0500BAEA
1785.81
CAD
H-5108.0500BA
H-5108.1000BA
OM99-2 Trifluoroacetate
Proteins and Peptides
OM99-2, EVNL-psi[CHOH-CH₂]-AAEF-NH₂, is a potent inhibitor for human brain memapsin-2 (β-secretase) containing a Leu-Ala hydroxyethylene isostere with a Ki-value of 9.58 · 10⁻⁹ M.