Human Adrenomedullin (13-52)
Supplier: BACHEM AMERICAS INC
4034489.0500 4034489.1000
H-4936.0500BAEA 1032.67 CAD
H-4936.0500BA H-4936.1000BA
Human Adrenomedullin (13-52)
Proteins and Peptides
This adrenomedullin fragment possessed vasodilatory activity. It has been suggested to modulate receptor-mediated, but not voltage-dependent, pulmonary vascular contraction by influencing Ca²⁺ influx. Therefore, it has been proposed that hADM (13-52) acts as an endothelium-dependent vasodilator agent in the pulmonary vascular bed of the rat. Furthermore, this compound may act as a modulator of the vascular phases of inflammation.